missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxybenzoic acid, 99%

Catalog No. p-206673
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC139930050 5 g Glass Bottle
AC139931000 100 g Plastic bottle
AC139935000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC139930050 Supplier Thermo Scientific Chemicals Supplier No. 139930050
Only null left

CAS: 118-41-2 | C10H12O5 | 212.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 118-41-2
Molecular Formula C10H12O5
Molecular Weight (g/mol) 212.2
MDL Number MFCD00002501
InChI Key SJSOFNCYXJUNBT-UHFFFAOYSA-N
Synonym eudesmic acid, gallic acid trimethyl ether, tri-o-methylgallic acid, trimethylgallic acid, benzoic acid, 3,4,5-trimethoxy, veratric acid, 5-methoxy, unii-v5c9h0sc9f, 3,4,5-trimethoxy-benzoic acid, 5-methoxy-veratric acid, v5c9h0sc9f
PubChem CID 8357
ChEBI CHEBI:454991
IUPAC Name 3,4,5-trimethoxybenzoic acid
SMILES COC1=CC(=CC(OC)=C1OC)C(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100
Melting Point 168.0°C to 172.0°C
Color Colorless to Yellow
Boiling Point 225.0°C to 227.0°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 0.99
Packaging Glass Bottle
Linear Formula (CH3O)3C6H2CO2H
Quantity 5 g
Beilstein 10, 481
Formula Weight 212.2
Percent Purity 0.99
Physical Form Liquid
Chemical Name or Material 3, 4, 5-Trimethoxybenzoic acid, 0.99
Show More Show Less
Hazard Statement GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 204-248-2
TSCA TSCA

RUO – Research Use Only