missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxybenzoic acid, 99+%

Catalog No. AAA1427422
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1427422 100 g
AAA1427436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1427422 Supplier Thermo Scientific Chemicals Supplier No. A1427422
Only null left

CAS: 118-41-2 | C10H12O5 | 212.2 g/mol

3,4,5-Trimethoxybenzoic acid is used as intermediates in medicine and organic synthesis. It acts as a metabolite of trimebutine. It is widely used in making dyes and inks, photographic developers and used as astringents in medically.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,5-Trimethoxybenzoic acid is used as intermediates in medicine and organic synthesis. It acts as a metabolite of trimebutine. It is widely used in making dyes and inks, photographic developers and used as astringents in medically.

Solubility
Soluble in ethanol, ethyl ether and chloroform. Slightly soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.

Chemical Identifiers

CAS 118-41-2
Molecular Formula C10H12O5
Molecular Weight (g/mol) 212.2
MDL Number MFCD00002501
InChI Key SJSOFNCYXJUNBT-UHFFFAOYSA-N
Synonym eudesmic acid, gallic acid trimethyl ether, tri-o-methylgallic acid, trimethylgallic acid, benzoic acid, 3,4,5-trimethoxy, veratric acid, 5-methoxy, unii-v5c9h0sc9f, 3,4,5-trimethoxy-benzoic acid, 5-methoxy-veratric acid, v5c9h0sc9f
PubChem CID 8357
ChEBI CHEBI:454991
IUPAC Name 3,4,5-trimethoxybenzoic acid
SMILES COC1=CC(=CC(OC)=C1OC)C(O)=O

Specifications

Melting Point 168°C to 171°C
Boiling Point 225°C to 226°C (10 mmHg)
Quantity 100 g
Beilstein 884655
Solubility Information Soluble in ethanol,ethyl ether and chloroform. Slightly soluble in water.
Formula Weight 212.2
Percent Purity ≥99%
Chemical Name or Material 3,4,5-Trimethoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 204-248-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only