Learn More
3,4,5-Trihydroxybenzoic acid monohydrate, ACS, 98+%
CAS: 5995-86-8 | C7H5O5 | 169.11 g/mol
$124.45 - $381.30
Chemical Identifiers
| Molecular Formula | C7H5O5 |
|---|---|
| Molecular Weight (g/mol) | 169.11 |
| MDL Number | MFCD00149098 |
| InChI Key | LNTHITQWFMADLM-UHFFFAOYSA-M |
| Synonym | gallic acid monohydrate, 3,4,5-trihydroxybenzoic acid hydrate, galop hydrate, 3,4,5-trihydroxybenzoic acid monohydrate, gallicum acidum, benzoic acid, 3,4,5-trihydroxy-, hydrate, gallic acid, monohydrate, gallicacidmonohydrate, gallic acid, acs, acmc-1ary0 |
| PubChem CID | 24721416 |
| SMILES | OC1=CC(=CC(O)=C1O)C([O-])=O |
Description
3,4,5-Trihydroxybenzoic acid monohydrate is used in making dyes and inks, photographic developers and has been used as astringents in medically. It is also are used as antioxidants in foods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C7H5O5 | |
| MFCD00149098 | |
| gallic acid monohydrate, 3,4,5-trihydroxybenzoic acid hydrate, galop hydrate, 3,4,5-trihydroxybenzoic acid monohydrate, gallicum acidum, benzoic acid, 3,4,5-trihydroxy-, hydrate, gallic acid, monohydrate, gallicacidmonohydrate, gallic acid, acs, acmc-1ary0 | |
| OC1=CC(=CC(O)=C1O)C([O-])=O |
| 169.11 | |
| LNTHITQWFMADLM-UHFFFAOYSA-M | |
| 24721416 |
Specifications
| 250°C (decomposition) | |
| C7H5O5 | |
| 100 g | |
| 144345 | |
| Soluble in water(1.1g/100ml). | |
| OC1=CC(=CC(O)=C1O)C([O-])=O | |
| 169.11 | |
| 188.14 (170.12 Anhydrous) | |
| ACS | |
| 3,4,5-Trihydroxybenzoic acid monohydrate |
| White to Cream | |
| MFCD00149098 | |
| Light sensitive | |
| gallic acid monohydrate, 3,4,5-trihydroxybenzoic acid hydrate, galop hydrate, 3,4,5-trihydroxybenzoic acid monohydrate, gallicum acidum, benzoic acid, 3,4,5-trihydroxy-, hydrate, gallic acid, monohydrate, gallicacidmonohydrate, gallic acid, acs, acmc-1ary0 | |
| LNTHITQWFMADLM-UHFFFAOYSA-M | |
| 3,4,5-trihydroxybenzoate | |
| 24721416 | |
| ≥98% | |
| Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 205-749-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only