missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Triethoxybenzoic acid, 98+%

Catalog No. AAA1415714
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1415714 25 g
AAA1415722 100 g
AAA1415736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1415714 Supplier Thermo Scientific Chemicals Supplier No. A1415714
Only null left

CAS: 6970-19-0 | C13H18O5 | 254.282 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6970-19-0
Molecular Formula C13H18O5
Molecular Weight (g/mol) 254.282
MDL Number MFCD00002505
InChI Key YCENSLDYFDZUMO-UHFFFAOYSA-N
Synonym benzoic acid,3,4,5-triethoxy, benzoic acid, 3,4,5-triethoxy, 3,4,5-triethoxy benzoic acid, acmc-1b720
PubChem CID 81432
IUPAC Name 3,4,5-triethoxybenzoic acid
SMILES CCOC1=CC(=CC(=C1OCC)OCC)C(=O)O

Specifications

Melting Point 110°C to 113°C
Quantity 25 g
Beilstein 2659063
Formula Weight 254.29
Percent Purity ≥98%
Chemical Name or Material 3,4,5-Triethoxybenzoic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 230-192-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only