missing translation for 'onlineSavingsMsg'
Learn More

3',4',5,7-Tetrahydroxyflavone 98.0+%, TCI America™

Catalog No. T26821G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26821G 1 g
T26825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26821G Supplier TCI America Supplier No. T26821G

Chemical Identifiers

CAS 491-70-3
Molecular Formula C15H10O6
Molecular Weight (g/mol) 286.239
MDL Number MFCD00017309
InChI Key IQPNAANSBPBGFQ-UHFFFAOYSA-N
Synonym luteolin, 3',4',5,7-tetrahydroxyflavone, 2-3,4-dihydroxyphenyl-5,7-dihydroxy-4h-chromen-4-one, digitoflavone, luteolol, flacitran, luteoline, weld lake, cyanidenon 1470, salifazide
PubChem CID 5280445
ChEBI CHEBI:15864
IUPAC Name 2-(3,4-dihydroxyphenyl)-5,7-dihydroxychromen-4-one
SMILES C1=CC(=C(C=C1C2=CC(=O)C3=C(C=C(C=C3O2)O)O)O)O

Specifications

Melting Point 330°C
Quantity 1 g
Formula Weight 286.24
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3′,4′,5,7-Tetrahydroxyflavone
RTECSNumber LK9275210
TSCA No
Recommended Storage Refrigerator