missing translation for 'onlineSavingsMsg'
Learn More

3',4',5,7-Tetrahydroxyflavone, 97%

Catalog No. AAL14186ME
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL14186ME 500 mg
AAL14186MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL14186ME Supplier Thermo Scientific Chemicals Supplier No. L14186ME

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 491-70-3 | C15H10O6 | 286.239 g/mol

3',4',5,7-Tetrahydroxyflavone is used as an apoptosis inducer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′,4′,5,7-Tetrahydroxyflavone is used as an apoptosis inducer.

Solubility
Soluble in aqueous alkaline solutions (1.4 mg/ml), ethanol (∼5 mg/ml), dimethyl sulfoxide (7 mg/ml), 1eq. Sodium hydroxide (5 mM), dimethylformamide (∼20 mg/ml), water (1 mg/ml) at 25°C and methanol.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C.

Chemical Identifiers

CAS 491-70-3
Molecular Formula C15H10O6
Molecular Weight (g/mol) 286.239
MDL Number MFCD00017309
InChI Key IQPNAANSBPBGFQ-UHFFFAOYSA-N
Synonym luteolin, 3',4',5,7-tetrahydroxyflavone, 2-3,4-dihydroxyphenyl-5,7-dihydroxy-4h-chromen-4-one, digitoflavone, luteolol, flacitran, luteoline, weld lake, cyanidenon 1470, salifazide
PubChem CID 5280445
ChEBI CHEBI:15864
IUPAC Name 2-(3,4-dihydroxyphenyl)-5,7-dihydroxychromen-4-one
SMILES C1=CC(=C(C=C1C2=CC(=O)C3=C(C=C(C=C3O2)O)O)O)O

Specifications

Melting Point >300°C
Quantity 500 mg
Beilstein 292084
Merck Index 14,5614
Solubility Information Soluble in aqueous alkaline solutions (1.4mg/ml),ethanol (∽5mg/ml),dimethyl sulfoxide (7mg/ml),1eq. Sodium hydroxide (5 mM),dimethylformamide (∽20mg/ml),water (1mg/ml) at 25°C and methanol.
Formula Weight 286.24
Percent Purity 97%
Chemical Name or Material 3',4',5,7-Tetrahydroxyflavone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-741-0
RTECSNumber LK9275210
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only