missing translation for 'onlineSavingsMsg'
Learn More

3,4,5,6-Tetrachloropyridine-2-carboxylic acid, 98%

Catalog No. p-7047563
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1945006 5 g
AAL1945014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1945006 Supplier Thermo Scientific Chemicals Supplier No. L1945006

CAS: 10469-09-7 | C6HCl4NO2 | 260.88 g/mol

3,4,5,6-Tetrachloropyridine-2-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceutical and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,5,6-Tetrachloropyridine-2-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceutical and dyestuff.

Solubility
Soluble in DMSO, Methanol. Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.

Chemical Identifiers

CAS 10469-09-7
Molecular Formula C6HCl4NO2
Molecular Weight (g/mol) 260.88
MDL Number MFCD00185833
InChI Key GXFRQLQUKBSYQL-UHFFFAOYSA-N
Synonym 3,4,5,6-tetrachloropicolinic acid, tetrachloropicolinic acid, tetrachloropyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 3,4,5,6-tetrachloro, unii-7e9zn8ab09, 3,4,5,6-tetrachloro-2-pyridinecarboxylic acid, 2-pyridinecarboxylicacid, 3,4,5,6-tetrachloro, tetrachloropyridine-2-carboxylicacid, dsstox_cid_24509, dsstox_rid_80279
PubChem CID 82646
IUPAC Name 3,4,5,6-tetrachloropyridine-2-carboxylic acid
SMILES OC(=O)C1=NC(Cl)=C(Cl)C(Cl)=C1Cl

Specifications

Melting Point 171°C to 173°C
Quantity 5 g
Beilstein 479080
Solubility Information Soluble in DMSO,Methanol. Insoluble in water.
Formula Weight 260.89
Percent Purity 98%
Chemical Name or Material 3,4,5,6-Tetrachloropyridine-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-956-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only