missing translation for 'onlineSavingsMsg'
Learn More

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98.0+%, TCI America™

Catalog No. p-7137168
Change view
Click to view available options
Quantity:
1 g
25 g
5 g
Catalog No. Quantity
T23451G 1 g
T23455G 5 g
T234525G 25 g
3 options

Catalog No. T23451G

Supplier: TCI America T23451G

Only null left

Chemical Identifiers

CAS 329214-79-1
Molecular Formula C11H16BNO2
Molecular Weight (g/mol) 205.06
MDL Number MFCD03084758
InChI Key XEMDFESAXKSEGI-UHFFFAOYSA-N
Synonym 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 3-pyridineboronic acid pinacol ester, pyridine-3-boronic acid pinacol ester, 3-pyridylboronic acid pinacol ester, 3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine-3-boronic acid, pinacol ester, 2-3-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, pyridine, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine
PubChem CID 2734656
IUPAC Name 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1(C)OB(OC1(C)C)C1=CC=CN=C1

Specifications

Melting Point 104°C
Color White-Yellow
Quantity 1 g
Formula Weight 205.06
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine