missing translation for 'onlineSavingsMsg'
Learn More

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic Acid 98.0+%, TCI America™

Catalog No. p-7137614
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31621G 1 g
T31625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31621G Supplier TCI America Supplier No. T31621G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 269409-73-6
Molecular Formula C13H17BO4
Molecular Weight (g/mol) 248.09
MDL Number MFCD03411930
InChI Key OPWAPCOSDAFWFB-UHFFFAOYSA-N
Synonym 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 3-carboxyphenylboronic acid pinacol ester, 3-carboxyphenylboronic acid, pinacol ester, 3-carboxyphenyl boronic acid, pinacol ester, benzoic acid, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 3-carboxyphenylboronicacidpinacolester, pubchem17048, 3-cpbe, amtb044
PubChem CID 2734653
IUPAC Name 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
SMILES CC1(C)OB(OC1(C)C)C1=CC=CC(=C1)C(O)=O

Specifications

Melting Point 210°C
Color White
Quantity 1 g
Formula Weight 248.09
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic Acid