Learn More
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 269409-73-6 |
|---|---|
| Molecular Formula | C13H17BO4 |
| Molecular Weight (g/mol) | 248.09 |
| MDL Number | MFCD03411930 |
| InChI Key | OPWAPCOSDAFWFB-UHFFFAOYSA-N |
| Synonym | 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 3-carboxyphenylboronic acid pinacol ester, 3-carboxyphenylboronic acid, pinacol ester, 3-carboxyphenyl boronic acid, pinacol ester, benzoic acid, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 3-carboxyphenylboronicacidpinacolester, pubchem17048, 3-cpbe, amtb044 |
| PubChem CID | 2734653 |
| IUPAC Name | 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid |
| SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC367570010
|
N/A | 1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC367570050
|
N/A | 5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 269409-73-6 | |
| 248.09 | |
| OPWAPCOSDAFWFB-UHFFFAOYSA-N | |
| 2734653 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C(=O)O |
| C13H17BO4 | |
| MFCD03411930 | |
| 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 3-carboxyphenylboronic acid pinacol ester, 3-carboxyphenylboronic acid, pinacol ester, 3-carboxyphenyl boronic acid, pinacol ester, benzoic acid, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 3-carboxyphenylboronicacidpinacolester, pubchem17048, 3-cpbe, amtb044 | |
| 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid |
Specifications
| 269409-73-6 | |
| 100.0 | |
| 97% | |
| MFCD03411930 | |
| Solubility in water: insoluble in water. | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C(=O)O | |
| 248.09 | |
| 248.09 | |
| 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 97% |
| 96.0 | |
| Authentic | |
| C13H17BO4 | |
| 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 3-carboxyphenylboronic acid pinacol ester, 3-carboxyphenylboronic acid, pinacol ester, 3-carboxyphenyl boronic acid, pinacol ester, benzoic acid, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 3-carboxyphenylboronicacidpinacolester, pubchem17048, 3-cpbe, amtb044 | |
| OPWAPCOSDAFWFB-UHFFFAOYSA-N | |
| 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid | |
| 2734653 | |
| 97% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
Recommended Storage : Refrigerator +4°C
RUO – Research Use Only