missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3-Tetramethyleneglutarimide 98.0+%, TCI America™
Supplier: TCI America T141210G
Specifications
| 3,3-Tetramethyleneglutarimide | |
| 154°C | |
| C9H13NO2 | |
| 10 g | |
| 3,3-tetramethyleneglutarimide, 8-azaspiro 4.5 decane-7,9-dione, azaspirodecanedione, 1,1-cyclopentanediacetimide, 8-aza-spiro 4.5 decane-7,9-dione, buspirone impurity k, dsstox_cid_31405, dsstox_rid_97291, dsstox_gsid_57616 | |
| C1CCC2(C1)CC(=O)NC(=O)C2 | |
| 167.208 | |
| 167.21 | |
| Crystalline Powder |
| 1075-89-4 | |
| White | |
| MFCD00023871 | |
| 2811 | |
| YRTHJMQKDCXPAY-UHFFFAOYSA-N | |
| 8-azaspiro[4.5]decane-7,9-dione | |
| 136843 | |
| ≥98.0% (HPLC,N) |
Chemical Identifiers
| 1075-89-4 | |
| 167.208 | |
| YRTHJMQKDCXPAY-UHFFFAOYSA-N | |
| 136843 | |
| C1CCC2(C1)CC(=O)NC(=O)C2 |
| C9H13NO2 | |
| MFCD00023871 | |
| 3,3-tetramethyleneglutarimide, 8-azaspiro 4.5 decane-7,9-dione, azaspirodecanedione, 1,1-cyclopentanediacetimide, 8-aza-spiro 4.5 decane-7,9-dione, buspirone impurity k, dsstox_cid_31405, dsstox_rid_97291, dsstox_gsid_57616 | |
| 8-azaspiro[4.5]decane-7,9-dione |
Safety and Handling
TSCA : No