missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Pyridyl)propionic Acid 98.0+%, TCI America™

Catalog No. P19921G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19921G 1 g
P19925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P19921G Supplier TCI America Supplier No. P19921G

Chemical Identifiers

CAS 3724-19-4
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00274197
InChI Key WDGXIUUWINKTGP-UHFFFAOYSA-N
Synonym 3-3-pyridyl propionic acid, 3-pyridinepropionic acid, 3-pyridinepropanoic acid, 3-pyridin-3-yl propanoic acid, 3-3-pyridyl propionicacid, pyridine-3-propionic acid, 3-3-pyridyl propanoic acid, 3-pyridin-3-yl propionic acid, pubchem8701, maybridge4_001332
PubChem CID 259624
IUPAC Name 3-pyridin-3-ylpropanoic acid
SMILES C1=CC(=CN=C1)CCC(=O)O

Specifications

Melting Point 158°C
Color White
Quantity 1 g
Formula Weight 151.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(3-Pyridyl)propionic Acid