missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Pyridyl)-L-alanine, 98%

Catalog No. AAH63941MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63941MD 250 mg
AAH6394103 1 g
AAH6394106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63941MD Supplier Thermo Scientific Chemicals Supplier No. H63941MD
Only null left

CAS: 64090-98-8 | C8H10N2O2 | 166.18 g/mol

3-(3-Pyridyl)-L-alanine is a antimycobacterial cyclodepsipeptide. PyrA is used to activate other aromatic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(3-Pyridyl)-L-alanine is a antimycobacterial cyclodepsipeptide. PyrA is used to activate other aromatic acids.

Solubility
Soluble in clear solution (0.3 gram in 2 ml DMF).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 64090-98-8
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00079676
InChI Key DFZVZEMNPGABKO-ZETCQYMHSA-N
Synonym 3-3-pyridyl-l-alanine, l-3-pyridylalanine, s-2-amino-3-pyridin-3-yl propanoic acid, h-3-pal-oh, 3-3-pyridyl alanine, 2s-2-amino-3-pyridin-3-yl propanoic acid, l-3-3-pyridyl-alanine, 3'-pyridyl-l-ala, h-beta-3-pyridyl-ala-oh, 2s-2-amino-3-pyridin-3-ylpropanoic acid
PubChem CID 152953
IUPAC Name (2S)-2-amino-3-pyridin-3-ylpropanoic acid
SMILES C1=CC(=CN=C1)CC(C(=O)O)N

Specifications

Melting Point 248°C to 252°C (decomposition)
Color White
Beilstein 4351583
Solubility Information Soluble in clear solution (0.3 gram in 2 ml DMF).
Quantity 250 mg
Formula Weight 166.18
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-(3-Pyridyl)-L-alanine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only