missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Piperidinyloxy)benzonitrile, 99%, Thermo Scientific Chemicals

$291.27 - $291.27

Chemical Identifiers

CAS 902836-93-5
Molecular Formula C12H14N2O
Molecular Weight (g/mol) 202.257
MDL Number MFCD08061079
InChI Key OSURJDMYHKQISX-UHFFFAOYSA-N
Synonym 3-3-piperidinyloxy benzonitrile, 3-piperidin-3-yloxy benzonitrile, 3-piperidin-3-yl-oxy benzonitrile, 3-3-piperidyloxy benzenecarbonitrile, 3-piperidin-3-yl oxy benzonitrile
PubChem CID 22309128
IUPAC Name 3-piperidin-3-yloxybenzonitrile
SMILES C1CC(CNC1)OC2=CC=CC(=C2)C#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5093903
View Documents
Thermo Scientific Chemicals
H5093903
1 g
Each for $291.27
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

902836-93-5
202.257
OSURJDMYHKQISX-UHFFFAOYSA-N
22309128
C1CC(CNC1)OC2=CC=CC(=C2)C#N
C12H14N2O
MFCD08061079
3-3-piperidinyloxy benzonitrile, 3-piperidin-3-yloxy benzonitrile, 3-piperidin-3-yl-oxy benzonitrile, 3-3-piperidyloxy benzenecarbonitrile, 3-piperidin-3-yl oxy benzonitrile
3-piperidin-3-yloxybenzonitrile

Specifications

902836-93-5
MFCD08061079
3-3-piperidinyloxy benzonitrile, 3-piperidin-3-yloxy benzonitrile, 3-piperidin-3-yl-oxy benzonitrile, 3-3-piperidyloxy benzenecarbonitrile, 3-piperidin-3-yl oxy benzonitrile
C1CC(CNC1)OC2=CC=CC(=C2)C#N
202.257
202.26
3-(3-Piperidinyloxy)benzonitrile
C12H14N2O
1 g
OSURJDMYHKQISX-UHFFFAOYSA-N
3-piperidin-3-yloxybenzonitrile
22309128
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only