missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Oxydipropionitrile 98.0+%, TCI America™

Catalog No. p-7118468
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B047525ML 25 mL
B0475500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B047525ML Supplier TCI America Supplier No. B047525ML

Chemical Identifiers

CAS 1656-48-0
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.143
MDL Number MFCD00001960
InChI Key BCGCCTGNWPKXJL-UHFFFAOYSA-N
Synonym bis 2-cyanoethyl ether, 2-cyanoethyl ether, 3,3'-oxydipropionitrile, propanenitrile, 3,3'-oxybis, ether, bis 2-cyanoethyl, 2,2'-dicyanodiethyl ether, propionitrile, 3,3'-oxydi, 3-2-cyanoethoxy propanenitrile, beta,beta'-dicyanodiethyl ether, unii-uf1uoo2pho
PubChem CID 15452
IUPAC Name 3-(2-cyanoethoxy)propanenitrile
SMILES C(COCCC#N)C#N

Specifications

Color Yellow
Boiling Point 172°C
Quantity 25 mL
Formula Weight 124.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,3′-Oxydipropionitrile
RTECSNumber UG4200000
TSCA Yes