missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Methyl-1,2,4-oxadiazol-5-yl)benzaldehyde, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC49504DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC49504DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

273727-50-7
188.19
YUNJEYQSCYYAEF-UHFFFAOYSA-N
18545510
CC1=NOC(=N1)C1=CC(C=O)=CC=C1
C10H8N2O2
MFCD09817468
3-3-methyl-1,2,4-oxadiazol-5-yl benzaldehyde, benzaldehyde,3-3-methyl-1,2,4-oxadiazol-5-yl, 3-3-methyl-1,2,4 oxadiazol-5-yl-benzaldehyde
3-(3-methyl-1,2,4-oxadiazol-5-yl)benzaldehyde

Specifications

3-(3-Methyl-1, 2, 4-oxadiazol-5-yl)benzaldehyde
96.5
97%
C10H8N2O2
MFCD09817468
YUNJEYQSCYYAEF-UHFFFAOYSA-N
3-(3-methyl-1,2,4-oxadiazol-5-yl)benzaldehyde
18545510
97%
273727-50-7
100.0
Amber Glass Bottle
1g
3-3-methyl-1,2,4-oxadiazol-5-yl benzaldehyde, benzaldehyde,3-3-methyl-1,2,4-oxadiazol-5-yl, 3-3-methyl-1,2,4 oxadiazol-5-yl-benzaldehyde
CC1=NOC(=N1)C1=CC(C=O)=CC=C1
188.19
188.19
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents