missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Methoxyphenyl)propionic acid, 98+%

Catalog No. p-7043561
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0130406 5 g
AAL0130414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0130406 Supplier Thermo Scientific Chemicals Supplier No. L0130406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10516-71-9 | C10H12O3 | 180.20 g/mol

3-(3-Methoxyphenyl)propionic acid is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(3-Methoxyphenyl)propionic acid is used as pharmaceutical intermediates.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 10516-71-9
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00014027
InChI Key BJJQJLOZWBZEGA-UHFFFAOYSA-N
Synonym 3-3-methoxyphenyl propionic acid, 3-3-methoxyphenyl propanoic acid, 3-m-methoxyphenyl propionic acid, 3-methoxybenzenepropanoic acid, benzenepropanoic acid, 3-methoxy, 3-3-methoxy-phenyl-propionic acid, 3-methoxy-benzenepropanoic acid, 3-3-methoxyphenyl propanoate, m-methoxyhydrocinnamate, 3-methoxydihydrocinnamate
PubChem CID 66336
IUPAC Name 3-(3-methoxyphenyl)propanoic acid
SMILES COC1=CC=CC(CCC(O)=O)=C1

Specifications

Melting Point 43°C to 47°C
Quantity 5 g
Beilstein 1874422
Solubility Information Soluble in water.
Formula Weight 180.2
Percent Purity ≥98%
Chemical Name or Material 3-(3-Methoxyphenyl)propionic acid
EINECSNumber 234-049-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only