missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Methoxyphenyl)propionic acid, 98+%

Catalog No. aal0130414
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
AAL0130414 25 g
AAL0130406 5 g
2 options

Catalog No. AAL0130414

Supplier: Thermo Scientific Chemicals L0130414

Only null left

CAS: 10516-71-9 | C10H12O3 | 180.20 g/mol

3-(3-Methoxyphenyl)propionic acid is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(3-Methoxyphenyl)propionic acid is used as pharmaceutical intermediates.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 10516-71-9
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00014027
InChI Key BJJQJLOZWBZEGA-UHFFFAOYSA-N
Synonym 3-3-methoxyphenyl propionic acid, 3-3-methoxyphenyl propanoic acid, 3-m-methoxyphenyl propionic acid, 3-methoxybenzenepropanoic acid, benzenepropanoic acid, 3-methoxy, 3-3-methoxy-phenyl-propionic acid, 3-methoxy-benzenepropanoic acid, 3-3-methoxyphenyl propanoate, m-methoxyhydrocinnamate, 3-methoxydihydrocinnamate
PubChem CID 66336
IUPAC Name 3-(3-methoxyphenyl)propanoic acid
SMILES COC1=CC=CC(CCC(O)=O)=C1

Specifications

Melting Point 43°C to 47°C
Quantity 25 g
Beilstein 1874422
Solubility Information Soluble in water.
Formula Weight 180.2
Percent Purity ≥98%
Chemical Name or Material 3-(3-Methoxyphenyl)propionic acid
EINECSNumber 234-049-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only