Learn More
Chemical Identifiers
| CAS | 6711-48-4 |
|---|---|
| Molecular Formula | C10H25N3 |
| Molecular Weight (g/mol) | 187.33 |
| InChI Key | BXYVQNNEFZOBOZ-UHFFFAOYSA-N |
| IUPAC Name | (3-{[3-(dimethylamino)propyl]amino}propyl)dimethylamine |
| SMILES | CN(C)CCCNCCCN(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC466340250
|
N/A | 25 mL |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC466341000
|
N/A | 100 mL |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC466345000
|
N/A | 500 mL |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 6711-48-4 | |
| 187.33 | |
| (3-{[3-(dimethylamino)propyl]amino}propyl)dimethylamine |
| C10H25N3 | |
| BXYVQNNEFZOBOZ-UHFFFAOYSA-N | |
| CN(C)CCCNCCCN(C)C |
Specifications
| 6711-48-4 | |
| 128.0°C to 131.0°C (27.0 hPa) | |
| C10H25N3 | |
| BXYVQNNEFZOBOZ-UHFFFAOYSA-N | |
| (3-{[3-(dimethylamino)propyl]amino}propyl)dimethylamine | |
| 187.33 |
| 0.8410 g/mL | |
| 98°C | |
| 0.841 | |
| CN(C)CCCNCCCN(C)C | |
| 187.33 | |
| 3,3 '-Iminobis(N,N-dimethylpropylamine) |
Safety and Handling
GHS H Statement Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water or shower IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
EINECSNumber : 229-761-9
RUO – Research Use Only