missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Iminobis(N,N-dimethylpropylamine) 97.0+%, TCI America™

Catalog No. I093925ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I093925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I093925ML Supplier TCI America Supplier No. I093925ML

Chemical Identifiers

CAS 6711-48-4
Molecular Formula C10H25N3
Molecular Weight (g/mol) 187.331
MDL Number MFCD00014880
InChI Key BXYVQNNEFZOBOZ-UHFFFAOYSA-N
Synonym N,N-Bis[3-(dimethylamino)propyl]amine, N′C-[3-(Dimethylamino)propyl]-N,N-dimethyl-1,3-propanediamine
PubChem CID 81207
IUPAC Name N-[3-(dimethylamino)propyl]-N',N'-dimethylpropane-1,3-diamine
SMILES CN(C)CCCNCCCN(C)C

Specifications

Color Colorless
Boiling Point 114°C
Quantity 25 mL
UN Number 2922
Formula Weight 187.33
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3,3′-Iminobis(N,N-dimethylpropylamine)
RTECSNumber JM1925000
TSCA Yes