missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Hydroxyphenyl)propionic acid, 98+%

Catalog No. aal0127914
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
AAL0127914 25 g
AAL0127906 5 g
2 options

Catalog No. AAL0127914

Supplier: Thermo Scientific Chemicals L0127914

Only null left

CAS: 621-54-5 | C9H10O3 | 166.176 g/mol

3-Hydroxyphenylpropionic acid acts as a urinary metabolite of procyanidins in pigs. It also serves as an intermediate in the preparation of various synthetic organic products. Further, it is used in the preparation of 3-(3-hydroxy-phenyl)-propionic acid methyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxyphenylpropionic acid acts as a urinary metabolite of procyanidins in pigs. It also serves as an intermediate in the preparation of various synthetic organic products. Further, it is used in the preparation of 3-(3-hydroxy-phenyl)-propionic acid methyl ester.

Solubility
Soluble in chloroform and methanol.

Notes
Incompatible with bases, oxidizing agents and reducing agents.

Chemical Identifiers

CAS 621-54-5
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00016554
InChI Key QVWAEZJXDYOKEH-UHFFFAOYSA-N
Synonym 3-3-hydroxyphenyl propanoic acid, 3-3-hydroxyphenyl propionic acid, dihydro-3-coumaric acid, benzenepropanoic acid, 3-hydroxy, 3-m-hydroxyphenyl propionic acid, 3-hydroxybenzenepropanoic acid, 3-hydroxyphenylpropionic acid, m-hydroxyphenylpropionic acid, beta-m-hydroxyphenyl propionic acid, hydrocinnamic acid, m-hydroxy
PubChem CID 91
ChEBI CHEBI:1427
IUPAC Name 3-(3-hydroxyphenyl)propanoic acid
SMILES C1=CC(=CC(=C1)O)CCC(=O)O

Specifications

Melting Point 110°C to 114°C
Quantity 25 g
Beilstein 1947445
Solubility Information Soluble in chloroform and methanol.
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material 3-(3-Hydroxyphenyl)propionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-692-8
RTECSNumber MW5340000
TSCA No

RUO – Research Use Only