missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(3-Furyl)acrylic acid, 98%
Description
3-(3-Furyl)acrylic acid is used as an intermediate in chemical research and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-(3-Furyl)acrylic acid is used as an intermediate in chemical research and pharmaceuticals.
Solubility
Sparingly soluble in water (0.56 g/L at 25°C).
Notes
Store at room temperature. Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
3-(3-Furyl)acrylic acid is used as an intermediate in chemical research and pharmaceuticals.
Solubility
Sparingly soluble in water (0.56 g/L at 25°C).
Notes
Store at room temperature. Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 152°C to 154°C |
| Quantity | 25 g |
| Beilstein | 1306450 |
| Solubility Information | Sparingly soluble in water (0.56g/L at 25°C). |
| Formula Weight | 138.12 |
| Percent Purity | 98% |
| Physical Form | Crystals or Powder or Crystalline Powder or Granules |
| Chemical Name or Material | 3-(3-Furyl)acrylic acid |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H StatementH315-H319-H335 Causes skin irritation.Causes serious eye irritation.May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only