missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Furyl)acrylic acid, 98%

Catalog No. AAL1314306
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1314306 5 g
AAL1314303 1 g
AAL1314314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1314306 Supplier Thermo Scientific Chemicals Supplier No. L1314306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 39244-10-5 | C7H6O3 | 138.12 g/mol

3-(3-Furyl)acrylic acid is used as an intermediate in chemical research and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(3-Furyl)acrylic acid is used as an intermediate in chemical research and pharmaceuticals.

Solubility
Sparingly soluble in water (0.56 g/L at 25°C).

Notes
Store at room temperature. Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 39244-10-5
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00075074
InChI Key JHAPZUDWRRBZHZ-UHFFFAOYSA-N
Synonym 3-3-furyl acrylic acid, 3-furan-3-yl acrylic acid, trans-3-furanacrylic acid, e-3-furan-3-yl acrylic acid, 2-propenoic acid, 3-3-furanyl-, 2e, 2e-3-3-furyl prop-2-enoic acid, 2e-3-furan-3-yl prop-2-enoic acid, 3-3-furanyl-2-propenoic acid, 2e-3-3-furyl acrylic acid, furan-3-acrylic acid
PubChem CID 643403
IUPAC Name (E)-3-(furan-3-yl)prop-2-enoic acid
SMILES OC(=O)C=CC1=COC=C1

Specifications

Melting Point 152°C to 154°C
Quantity 5 g
Beilstein 1306450
Solubility Information Sparingly soluble in water (0.56g/L at 25°C).
Formula Weight 138.12
Percent Purity 98%
Physical Form Crystals or Powder or Crystalline Powder or Granules
Chemical Name or Material 3-(3-Furyl)acrylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement

H315-H319-H335

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only