missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Dipropylthiadicarbocyanine Iodide 98.0+%, TCI America™

Catalog No. p-7128284
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4456200MG 200 mg
D44561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4456200MG Supplier TCI America Supplier No. D4456200MG
Only null left

Chemical Identifiers

CAS 53213-94-8
Molecular Formula C25H27IN2S2
Molecular Weight (g/mol) 546.53
MDL Number MFCD00075033,MFCD00075033
InChI Key GDEURKKLNUGTDA-UHFFFAOYSA-M
Synonym disc3 5, 3,3'-dipropylthiadicarbocyanine iodide, 3-propyl-2-5-3-propyl-2 3h-benzothiazolylidene-1,3-pentadienyl benzothiazolium iodide, 5-3-propylbenzothiazol-2-ylidene-1-3-propylbenzothiazolium-2-yl-1,3-pentadiene iodide
PubChem CID 91972132
IUPAC Name 3-propyl-2-[(3E)-5-[(2Z)-3-propyl-2,3-dihydro-1,3-benzothiazol-2-ylidene]penta-1,3-dien-1-yl]-1,3-benzothiazol-3-ium iodide
SMILES [I-].CCCN1\C(SC2=CC=CC=C12)=C\C=C\C=CC1=[N+](CCC)C2=CC=CC=C2S1

Specifications

Melting Point 245°C
Quantity 200 mg
Formula Weight 546.53
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Dipropylthiadicarbocyanine Iodide