missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3'-Dipropylthiadicarbocyanine Iodide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 245°C |
| Quantity | 200 mg |
| Formula Weight | 546.53 |
| Percent Purity | ≥98.0% (N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3,3′-Dipropylthiadicarbocyanine Iodide |
Safety and Handling
| TSCA | No |