missing translation for 'onlineSavingsMsg'
Learn More

3,3-Diphenylpropionitrile, Thermo Scientific Chemicals

Catalog No. AC466080250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466080250 25 g
AC466080050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466080250 Supplier Thermo Scientific Chemicals Supplier No. 466080250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2286-54-6
MDL Number MFCD00129747
InChI Key INERKLNEVAZSCI-UHFFFAOYSA-N
IUPAC Name 3,3-diphenylpropanenitrile
SMILES N#CCC(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Alpha Vector DIPHENYLPROPIONITRILE 010
Melting Point 86°C to 89°C
Color White to Yellow
Infrared Spectrum Conforms
Packaging glass bottle
Molecular Formula C15H13N
Quantity 25 g
Solubility Information soluble in hot methanol
Molecular Weight (g/mol) 207.28
Formula Weight 207.27
Physical Form Crystalline Powder
Chemical Name or Material 3,3-Diphenylpropionitrile
Show More Show Less
Hazard Category GHS07: Harmful
ShelfLife 5 years
storageNote1 normal conditions

RUO – Research Use Only