missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylpentane, 99%

Catalog No. p-3778413
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC152660010 1 g
AC152660050 5 g
AC152660250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC152660010 Supplier Thermo Scientific Chemicals Supplier No. 152660010

CAS: 562-49-2 | C7H16 | 100.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 562-49-2
Molecular Formula C7H16
Molecular Weight (g/mol) 100.2
MDL Number MFCD00009322
InChI Key AEXMKKGTQYQZCS-UHFFFAOYSA-N
PubChem CID 11229
IUPAC Name 3,3-dimethylpentane
SMILES CCC(C)(C)CC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -135°C
Color Colorless
Density 0.6900g/mL
Boiling Point 86°C
Flash Point -6°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.3900 to 1.392
Linear Formula C2H5C(CH3)2C2H5
Quantity 1 g
Beilstein 01,158
Specific Gravity 0.69
Synonym pentane, 3,3-dimethyl, pentane,3,3-dimethyl, unii-r1rh0vbd2l, r1rh0vbd2l, 3,3,-dimethylpentane, acmc-1b14l, dsstox_cid_29275, dsstox_rid_83394, dsstox_gsid_49319, 3,3-dimethylpentane
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,ethyl ether,miscible with acetone
Formula Weight 100.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3, 3-Dimethylpentane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-230-8

RUO – Research Use Only