missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylglutaric Anhydride 97.0+%, TCI America™

Catalog No. D13235G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D13235G 5 g
D132325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D13235G Supplier TCI America Supplier No. D13235G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4160-82-1
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.154
MDL Number MFCD00006684
InChI Key HIJQFTSZBHDYKW-UHFFFAOYSA-N
Synonym 3,3-dimethylglutaric anhydride, 2h-pyran-2,6 3h-dione, dihydro-4,4-dimethyl, 4,4-dimethyldihydro-2h-pyran-2,6 3h-dione, glutaric anhydride, 3,3-dimethyl, 3,3-dimethylglutaricanhydride, glutaric anhydride, beta,beta-dimethyl, acmc-1adve, dihydro-4,4-dimethyl-2h-pyran-2,6 3h-dione, 3,3-dimethyladipic anhydride, 3,3 dimethylglutaric anhydride
PubChem CID 77813
IUPAC Name 4,4-dimethyloxane-2,6-dione
SMILES CC1(CC(=O)OC(=O)C1)C

Specifications

Melting Point 126°C
Color White
Quantity 5 g
Formula Weight 142.15
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3-Dimethylglutaric Anhydride