missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylglutaric anhydride, 97%

Catalog No. AAB2528614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2528614 25 g
AAB2528606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2528614 Supplier Thermo Scientific Chemicals Supplier No. B2528614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 4160-82-1 | C7H10O3 | 142.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4160-82-1
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.154
MDL Number MFCD00006684
InChI Key HIJQFTSZBHDYKW-UHFFFAOYSA-N
Synonym 3,3-dimethylglutaric anhydride, 2h-pyran-2,6 3h-dione, dihydro-4,4-dimethyl, 4,4-dimethyldihydro-2h-pyran-2,6 3h-dione, glutaric anhydride, 3,3-dimethyl, 3,3-dimethylglutaricanhydride, glutaric anhydride, beta,beta-dimethyl, acmc-1adve, dihydro-4,4-dimethyl-2h-pyran-2,6 3h-dione, 3,3-dimethyladipic anhydride, 3,3 dimethylglutaric anhydride
PubChem CID 77813
IUPAC Name 4,4-dimethyloxane-2,6-dione
SMILES CC1(CC(=O)OC(=O)C1)C

Specifications

Melting Point 122°C to 126°C
Boiling Point 181°C (25 mmHg)
Quantity 25 g
Sensitivity Moisture sensitive
Formula Weight 142.15
Percent Purity 97%
Chemical Name or Material 3,3-Dimethylglutaric anhydride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 223-998-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only