missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylglutaric Acid 98.0+%, TCI America™

Catalog No. D132225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D132225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D132225G Supplier TCI America Supplier No. D132225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4839-46-7
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.17
MDL Number MFCD00002716
InChI Key DUHQIGLHYXLKAE-UHFFFAOYSA-N
Synonym 3,3-dimethylglutaric acid, pentanedioic acid, 3,3-dimethyl, 3,3-dimethylglutarate, 3,3-dimethyl-pentanedioic acid, .beta.,.beta.-dimethylglutaric acid, 3,3-dimethyl-glutaric acid, 3,3-dimethylpentaneodioic acid, glutaric acid,3-dimethyl, pentanedioic acid,3-dimethyl, beta,beta-dimethylglutaric acid
PubChem CID 20984
ChEBI CHEBI:68502
IUPAC Name 3,3-dimethylpentanedioic acid
SMILES CC(C)(CC(O)=O)CC(O)=O

Specifications

Melting Point 103°C
Color White-Yellow
Quantity 25 g
Formula Weight 160.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3-Dimethylglutaric Acid