missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3-Dimethylglutaric acid, 98%
CAS: 4839-46-7 | C7H12O4 | 160.17 g/mol
$182.67 - $182.67
Chemical Identifiers
| CAS | 4839-46-7 |
|---|---|
| Molecular Formula | C7H12O4 |
| Molecular Weight (g/mol) | 160.17 |
| MDL Number | MFCD00002716 |
| InChI Key | DUHQIGLHYXLKAE-UHFFFAOYSA-N |
| Synonym | 3,3-dimethylglutaric acid, pentanedioic acid, 3,3-dimethyl, 3,3-dimethylglutarate, 3,3-dimethyl-pentanedioic acid, .beta.,.beta.-dimethylglutaric acid, 3,3-dimethyl-glutaric acid, 3,3-dimethylpentaneodioic acid, glutaric acid,3-dimethyl, pentanedioic acid,3-dimethyl, beta,beta-dimethylglutaric acid |
| PubChem CID | 20984 |
| ChEBI | CHEBI:68502 |
| IUPAC Name | 3,3-dimethylpentanedioic acid |
| SMILES | CC(C)(CC(O)=O)CC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC116240250
|
Thermo Scientific Chemicals
116240250 |
25 g | Glass bottle |
Each for $182.67
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 4839-46-7 | |
| 160.17 | |
| DUHQIGLHYXLKAE-UHFFFAOYSA-N | |
| 20984 | |
| 3,3-dimethylpentanedioic acid |
| C7H12O4 | |
| MFCD00002716 | |
| 3,3-dimethylglutaric acid, pentanedioic acid, 3,3-dimethyl, 3,3-dimethylglutarate, 3,3-dimethyl-pentanedioic acid, .beta.,.beta.-dimethylglutaric acid, 3,3-dimethyl-glutaric acid, 3,3-dimethylpentaneodioic acid, glutaric acid,3-dimethyl, pentanedioic acid,3-dimethyl, beta,beta-dimethylglutaric acid | |
| CHEBI:68502 | |
| CC(C)(CC(O)=O)CC(O)=O |
Specifications
| 4839-46-7 | |
| 100.0 | |
| White to Beige | |
| Authentic | |
| Glass bottle | |
| HO2CCH2C(CH3)2CH2CO2H | |
| 25 g | |
| 3,3-dimethylglutaric acid, pentanedioic acid, 3,3-dimethyl, 3,3-dimethylglutarate, 3,3-dimethyl-pentanedioic acid, .beta.,.beta.-dimethylglutaric acid, 3,3-dimethyl-glutaric acid, 3,3-dimethylpentaneodioic acid, glutaric acid,3-dimethyl, pentanedioic acid,3-dimethyl, beta,beta-dimethylglutaric acid | |
| DUHQIGLHYXLKAE-UHFFFAOYSA-N | |
| 3,3-dimethylpentanedioic acid | |
| 20984 | |
| 160.17 | |
| Fine Crystalline Powder |
| 97.5 | |
| 98.0°C to 102.0°C | |
| 290.0°C | |
| 98% | |
| C7H12O4 | |
| MFCD00002716 | |
| 02, 684 | |
| Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol and ether,sparingly soluble in benzene and chloroform,insoluble in low-boiling petroleum fractions | |
| CC(C)(CC(O)=O)CC(O)=O | |
| 160.17 | |
| CHEBI:68502 | |
| 98% | |
| 3, 3-Dimethylglutaric acid |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 225-425-
RUO – Research Use Only