missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3-Dimethylcyclohexanone 97.0+%, TCI America™
$50.33 - $150.98
Chemical Identifiers
| CAS | 2979-19-3 |
|---|---|
| Molecular Formula | C8H14O |
| Molecular Weight (g/mol) | 126.20 |
| MDL Number | MFCD00040178 |
| InChI Key | ZVJQBBYAVPAFLX-UHFFFAOYSA-N |
| Synonym | 3,3-dimethylcyclohexanone, cyclohexanone, 3,3-dimethyl, 3,3-dimethylcyclohexane-1-one, 3,3-dimethyl-cyclohexanone, 3,3-dimethylcyclohexanon, acmc-209hb2, 3,3-dimethyl cyclohexanone, ksc497k3f, 3,3-dimethyl-cyclohexan-1-one, zvjqbbyavpaflx-uhfffaoysa |
| PubChem CID | 76322 |
| IUPAC Name | 3,3-dimethylcyclohexan-1-one |
| SMILES | CC1(C)CCCC(=O)C1 |
Chemical Identifiers
| 2979-19-3 | |
| 126.20 | |
| ZVJQBBYAVPAFLX-UHFFFAOYSA-N | |
| 76322 | |
| CC1(C)CCCC(=O)C1 |
| C8H14O | |
| MFCD00040178 | |
| 3,3-dimethylcyclohexanone, cyclohexanone, 3,3-dimethyl, 3,3-dimethylcyclohexane-1-one, 3,3-dimethyl-cyclohexanone, 3,3-dimethylcyclohexanon, acmc-209hb2, 3,3-dimethyl cyclohexanone, ksc497k3f, 3,3-dimethyl-cyclohexan-1-one, zvjqbbyavpaflx-uhfffaoysa | |
| 3,3-dimethylcyclohexan-1-one |
Specifications
| 2979-19-3 | |
| 175°C | |
| MFCD00040178 | |
| 1224 | |
| ZVJQBBYAVPAFLX-UHFFFAOYSA-N | |
| 3,3-dimethylcyclohexan-1-one | |
| 76322 | |
| ≥97.0% (GC) | |
| 3,3-Dimethylcyclohexanone |
| Yellow | |
| C8H14O | |
| 1 g | |
| 3,3-dimethylcyclohexanone, cyclohexanone, 3,3-dimethyl, 3,3-dimethylcyclohexane-1-one, 3,3-dimethyl-cyclohexanone, 3,3-dimethylcyclohexanon, acmc-209hb2, 3,3-dimethyl cyclohexanone, ksc497k3f, 3,3-dimethyl-cyclohexan-1-one, zvjqbbyavpaflx-uhfffaoysa | |
| CC1(C)CCCC(=O)C1 | |
| 126.20 | |
| 126.20 | |
| Liquid |
Safety and Handling
TSCA : Yes