missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3-Dimethylcyclohexanone 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 175°C |
| Quantity | 1 g |
| UN Number | 1224 |
| Formula Weight | 126.20 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 3,3-Dimethylcyclohexanone |
Safety and Handling
| TSCA | Yes |