Learn More
3,3-Dimethylcyclohexanone, 97%, Thermo Scientific Chemicals
$170.00 - $170.00
Chemical Identifiers
| CAS | 2979-19-3 |
|---|---|
| Molecular Formula | C8H14O |
| Molecular Weight (g/mol) | 126.20 |
| MDL Number | MFCD00040178 |
| InChI Key | ZVJQBBYAVPAFLX-UHFFFAOYSA-N |
| Synonym | 3,3-dimethylcyclohexanone, cyclohexanone, 3,3-dimethyl, 3,3-dimethylcyclohexane-1-one, 3,3-dimethyl-cyclohexanone, 3,3-dimethylcyclohexanon, acmc-209hb2, 3,3-dimethyl cyclohexanone, ksc497k3f, 3,3-dimethyl-cyclohexan-1-one, zvjqbbyavpaflx-uhfffaoysa |
| PubChem CID | 76322 |
| IUPAC Name | 3,3-dimethylcyclohexan-1-one |
| SMILES | CC1(C)CCCC(=O)C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2800006
|
Thermo Scientific Chemicals
H2800006 |
5 g |
Each for $170.00
|
|
|||||
Description
An interest in the boll weevil sex attractants prompted an investigation into alternative syntheses of the starting material, 3, 3-dimethylcyclohexanone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAn interest in the boll weevil sex attractants prompted an investigation into alternative syntheses of the starting material, 3, 3-dimethylcyclohexanone.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
| 2979-19-3 | |
| 126.20 | |
| ZVJQBBYAVPAFLX-UHFFFAOYSA-N | |
| 76322 | |
| CC1(C)CCCC(=O)C1 |
| C8H14O | |
| MFCD00040178 | |
| 3,3-dimethylcyclohexanone, cyclohexanone, 3,3-dimethyl, 3,3-dimethylcyclohexane-1-one, 3,3-dimethyl-cyclohexanone, 3,3-dimethylcyclohexanon, acmc-209hb2, 3,3-dimethyl cyclohexanone, ksc497k3f, 3,3-dimethyl-cyclohexan-1-one, zvjqbbyavpaflx-uhfffaoysa | |
| 3,3-dimethylcyclohexan-1-one |
Specifications
| 2979-19-3 | |
| 175°C | |
| C8H14O | |
| MFCD00040178 | |
| UN1224 | |
| Insoluble in water. | |
| CC1(C)CCCC(=O)C1 | |
| 126.20 | |
| 126.2 | |
| 3,3-Dimethylcyclohexanone |
| 0.91 | |
| 59°C (138°F) | |
| 1.447 | |
| 5 g | |
| 3,3-dimethylcyclohexanone, cyclohexanone, 3,3-dimethyl, 3,3-dimethylcyclohexane-1-one, 3,3-dimethyl-cyclohexanone, 3,3-dimethylcyclohexanon, acmc-209hb2, 3,3-dimethyl cyclohexanone, ksc497k3f, 3,3-dimethyl-cyclohexan-1-one, zvjqbbyavpaflx-uhfffaoysa | |
| ZVJQBBYAVPAFLX-UHFFFAOYSA-N | |
| 3,3-dimethylcyclohexan-1-one | |
| 76322 | |
| 97% |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only