missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylbutyraldehyde 97.0+%, TCI America™

Catalog No. D40985ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40985ML 5 mL
D409825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40985ML Supplier TCI America Supplier No. D40985ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2987-16-8
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00042807
InChI Key LTNUSYNQZJZUSY-UHFFFAOYSA-N
Synonym 3,3-dimethylbutyraldehyde, 3,3-dimethyl-butyraldehyde, butanal, 3,3-dimethyl, unii-cam6hd7jki, tert-butylacetaldehyde, 3,3-dimethyl-butanal, cam6hd7jki, neohexal, pubchem3076, 3,3-dimethylbutyraldehyd
PubChem CID 76335
IUPAC Name 3,3-dimethylbutanal
SMILES CC(C)(C)CC=O

Specifications

Color Colorless
Boiling Point 104°C
Quantity 5 mL
UN Number 1989
Formula Weight 100.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,3-Dimethylbutyraldehyde
EINECSNumber (2)-0494
TSCA No