missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylbutyraldehyde, 97%

Catalog No. p-7037003
Change view
Click to view available options
Quantity:
1 mL
5 mL
Catalog No. Quantity
AAH55522XF 1 mL
AAH55522AA 5 mL
2 options

Catalog No. AAH55522XF

Supplier: Thermo Scientific Chemicals H55522XF

Only null left

CAS: 2987-16-8 | C6H12O | 100.161 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2987-16-8
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00042807
InChI Key LTNUSYNQZJZUSY-UHFFFAOYSA-N
Synonym 3,3-dimethylbutyraldehyde, 3,3-dimethyl-butyraldehyde, butanal, 3,3-dimethyl, unii-cam6hd7jki, tert-butylacetaldehyde, 3,3-dimethyl-butanal, cam6hd7jki, neohexal, pubchem3076, 3,3-dimethylbutyraldehyd
PubChem CID 76335
IUPAC Name 3,3-dimethylbutanal
SMILES CC(C)(C)CC=O

Specifications

Density 0.798
Boiling Point 104°C to 106°C
Flash Point 13°C (55°F)
Refractive Index 1.397
Linear Formula (CH3)3CCH2CHO
Quantity 1 mL
UN Number UN1989
Beilstein 1735829
Sensitivity Air and moisture sensitive
Formula Weight 100.16
Percent Purity 97%
Chemical Name or Material 3,3-Dimethylbutyraldehyde
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 221-054-3
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only