missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylbutylamine, 95%, Thermo Scientific™

Catalog No. p-3593270
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC179380010 1g Glass bottle
AC179380050 5g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC179380010 Supplier Thermo Scientific Chemicals Supplier No. 179380010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 15673-00-4
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
MDL Number MFCD00008201
InChI Key GPWHFPWZAPOYNO-UHFFFAOYSA-N
Synonym 3,3-dimethylbutylamine, 1-butanamine, 3,3-dimethyl, 3,3-dimethyl butylamine, 3,3-dimethyl-butylamine, 3,3-dimethylbutyamine, 3,3 dimethylbutylamine, acmc-1btoo, 3,3-dimethylbutyl amine, 1-butanamine,3,3-dimethyl
PubChem CID 19709
IUPAC Name 3,3-dimethylbutan-1-amine
SMILES CC(C)(C)CCN

Specifications

CAS Min % 94.0
CAS Max % 100.0
Density 0.7520g/mL
Boiling Point 114°C to 116°C
Flash Point 6°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4120 to 1.4160 (20°C, 589nm)
Linear Formula (CH3)3CCH2CH2NH2
Quantity 1g
Specific Gravity 0.752
Formula Weight 101.19
Percent Purity 95%
Chemical Name or Material 3, 3-Dimethylbutylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 239-749-5

RUO – Research Use Only