missing translation for 'onlineSavingsMsg'
Learn More

{3-[3-(Dimethylamino)propoxy]phenyl}methanol, 97%, Thermo Scientific™

Catalog No. CC59409DE
Change view
Click to view available options
Quantity:
5g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC59409DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC59409DE Supplier Thermo Scientific Chemicals Supplier No. CC59409DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 912569-56-3
Molecular Formula C12H19NO2
Molecular Weight (g/mol) 209.289
MDL Number MFCD09064989
InChI Key BABQTNDHVRVEJD-UHFFFAOYSA-N
Synonym 3-3-dimethylamino propoxy phenyl methanol, benzenemethanol, 3-3-dimethylamino propoxy, 3-3-dimethylamino-propoxy-phenyl-methanol, 3-3-dimethylamino propoxy phenyl methan-1-ol
PubChem CID 24229604
IUPAC Name [3-[3-(dimethylamino)propoxy]phenyl]methanol
SMILES CN(C)CCCOC1=CC=CC(=C1)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5g
Formula Weight 209.29
Percent Purity 97%
Chemical Name or Material {3-[3-(Dimethylamino)propoxy]phenyl}methanol
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.