Learn More
3,3-Dimethylallyl acetate, 98%, stab. with 0.1% alpha tocopherol, Thermo Scientific Chemicals
$89.15 - $89.15
Chemical Identifiers
| CAS | 1191-16-8 |
|---|---|
| Molecular Formula | C7H12O2 |
| Molecular Weight (g/mol) | 128.17 |
| MDL Number | MFCD00036569 |
| InChI Key | XXIKYCPRDXIMQM-UHFFFAOYSA-N |
| Synonym | prenyl acetate, 3,3-dimethylallyl acetate, 3-methyl-2-butenyl acetate, isopent-2-enyl acetate, 3-methylbut-2-en-1-yl acetate, dimethylallyl acetate, 2-buten-1-ol, 3-methyl-, acetate, 3-methyl-2-buten-1-ol, acetate, unii-i7kov03hgs, isopentenyl acetate |
| PubChem CID | 14489 |
| IUPAC Name | 3-methylbut-2-enyl acetate |
| SMILES | CC(C)=CCOC(C)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1895818
|
Thermo Scientific Chemicals
A1895818 |
50 g |
Each for $89.15
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1191-16-8 | |
| 128.17 | |
| XXIKYCPRDXIMQM-UHFFFAOYSA-N | |
| 14489 | |
| CC(C)=CCOC(C)=O |
| C7H12O2 | |
| MFCD00036569 | |
| prenyl acetate, 3,3-dimethylallyl acetate, 3-methyl-2-butenyl acetate, isopent-2-enyl acetate, 3-methylbut-2-en-1-yl acetate, dimethylallyl acetate, 2-buten-1-ol, 3-methyl-, acetate, 3-methyl-2-buten-1-ol, acetate, unii-i7kov03hgs, isopentenyl acetate | |
| 3-methylbut-2-enyl acetate |
Specifications
| 1191-16-8 | |
| 140°C | |
| Fruit-like | |
| 1.43 | |
| 50 g | |
| prenyl acetate, 3,3-dimethylallyl acetate, 3-methyl-2-butenyl acetate, isopent-2-enyl acetate, 3-methylbut-2-en-1-yl acetate, dimethylallyl acetate, 2-buten-1-ol, 3-methyl-, acetate, 3-methyl-2-buten-1-ol, acetate, unii-i7kov03hgs, isopentenyl acetate | |
| CC(C)=CCOC(C)=O | |
| 128.17 | |
| 128.17 | |
| 3,3-Dimethylallyl acetate, stabilized with 0.1% alpha tocopherol |
| 0.915 | |
| 40°C (104°F) | |
| C7H12O2 | |
| MFCD00036569 | |
| UN3272 | |
| XXIKYCPRDXIMQM-UHFFFAOYSA-N | |
| 3-methylbut-2-enyl acetate | |
| 14489 | |
| 98% |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 214-730-4
RTECSNumber : EM9473700
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only