missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylallyl acetate, 98%, stab. with 0.1% alpha tocopherol, Thermo Scientific Chemicals

Catalog No. AAA1895818
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1895818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1895818 Supplier Thermo Scientific Chemicals Supplier No. A1895818

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1191-16-8
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00036569
InChI Key XXIKYCPRDXIMQM-UHFFFAOYSA-N
Synonym prenyl acetate, 3,3-dimethylallyl acetate, 3-methyl-2-butenyl acetate, isopent-2-enyl acetate, 3-methylbut-2-en-1-yl acetate, dimethylallyl acetate, 2-buten-1-ol, 3-methyl-, acetate, 3-methyl-2-buten-1-ol, acetate, unii-i7kov03hgs, isopentenyl acetate
PubChem CID 14489
IUPAC Name 3-methylbut-2-enyl acetate
SMILES CC(C)=CCOC(C)=O

Specifications

Density 0.915
Boiling Point 140°C
Flash Point 40°C (104°F)
Odor Fruit-like
Refractive Index 1.43
Quantity 50 g
UN Number UN3272
Formula Weight 128.17
Percent Purity 98%
Chemical Name or Material 3,3-Dimethylallyl acetate, stabilized with 0.1% alpha tocopherol
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 214-730-4
RTECSNumber EM9473700
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only