missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylacryloyl chloride, 97%

CAS: 3350-78-5 | C5H7ClO | 118.56 g/mol

$342.14 - $342.14

Chemical Identifiers

CAS 3350-78-5
Molecular Formula C5H7ClO
Molecular Weight (g/mol) 118.56
MDL Number MFCD00000730
InChI Key BDUBTLFQHNYXPC-UHFFFAOYSA-N
Synonym 3,3-dimethylacryloyl chloride, 3-methylcrotonoyl chloride, 3-methyl-but-2-enoyl chloride, 3,3-dimethylacrylyl chloride, 2-butenoyl chloride, 3-methyl, 3-methyl-2-butenoyl chloride, 3,3-dimethylacryloylchloride, dimethylacrylyl chloride, dimethyl acryloyl chloride, acmc-1cpb0
PubChem CID 102394
IUPAC Name 3-methylbut-2-enoyl chloride
SMILES CC(C)=CC(Cl)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC179730250
SDS View Documents
Thermo Scientific Chemicals
179730250
25 g Glass bottle
Each for $342.14
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3350-78-5
118.56
BDUBTLFQHNYXPC-UHFFFAOYSA-N
102394
CC(C)=CC(Cl)=O
C5H7ClO
MFCD00000730
3,3-dimethylacryloyl chloride, 3-methylcrotonoyl chloride, 3-methyl-but-2-enoyl chloride, 3,3-dimethylacrylyl chloride, 2-butenoyl chloride, 3-methyl, 3-methyl-2-butenoyl chloride, 3,3-dimethylacryloylchloride, dimethylacrylyl chloride, dimethyl acryloyl chloride, acmc-1cpb0
3-methylbut-2-enoyl chloride

Specifications

3350-78-5
100.0
1.0600g/mL
51°C
96% min. (GC)
C5H7ClO
(CH3)2C=CHCOCl
25 g
1.06
Solubility in water: reacts
CC(C)=CC(Cl)=O
118.56
118.56
Liquid
96.0
Colorless to Yellow
145.0°C to 147.0°C
Authentic
Glass bottle
1.4750 to 1.4780
MFCD00000730
02, 433
3,3-dimethylacryloyl chloride, 3-methylcrotonoyl chloride, 3-methyl-but-2-enoyl chloride, 3,3-dimethylacrylyl chloride, 2-butenoyl chloride, 3-methyl, 3-methyl-2-butenoyl chloride, 3,3-dimethylacryloylchloride, dimethylacrylyl chloride, dimethyl acryloyl chloride, acmc-1cpb0
BDUBTLFQHNYXPC-UHFFFAOYSA-N
3-methylbut-2-enoyl chloride
102394
97%
3, 3-Dimethylacryloyl chloride, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 222-109-4

TSCA : TSCA

Recommended Storage : Product may darken on storage

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only