Learn More
3,3-Dimethylacrylic acid, 98%
CAS: 541-47-9 | C5H8O2 | 100.117 g/mol
Supplier: Thermo Scientific Chemicals A1686522
Description
Chemical Intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsChemical Intermediates.
Solubility
Soluble in water, and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from bases.
Specifications
| 3,3-Dimethylacrylic acid | |
| 66°C to 68°C | |
| 198°C to 200°C | |
| C5H8O2 | |
| 100 g | |
| 1720305 | |
| Soluble in water,and methanol. | |
| CC(=CC(=O)O)C | |
| 100.117 | |
| CHEBI:37127 | |
| 98% |
| 541-47-9 | |
| 0.969 | |
| 93°C (199°F) | |
| MFCD00004366 | |
| UN3261 | |
| 3,3-dimethylacrylic acid, senecioic acid, 3-methylcrotonic acid, 3-methyl-2-butenoic acid, 2-butenoic acid, 3-methyl, senecic acid, beta,beta-dimethylacrylic acid, crotonic acid, 3-methyl, beta,beta-dimethacrylic acid, beta-methylcrotonic acid | |
| YYPNJNDODFVZLE-UHFFFAOYSA-N | |
| 3-methylbut-2-enoic acid | |
| 10931 | |
| 100.12 |
Chemical Identifiers
| 541-47-9 | |
| 100.117 | |
| YYPNJNDODFVZLE-UHFFFAOYSA-N | |
| 10931 | |
| 3-methylbut-2-enoic acid |
| C5H8O2 | |
| MFCD00004366 | |
| 3,3-dimethylacrylic acid, senecioic acid, 3-methylcrotonic acid, 3-methyl-2-butenoic acid, 2-butenoic acid, 3-methyl, senecic acid, beta,beta-dimethylacrylic acid, crotonic acid, 3-methyl, beta,beta-dimethacrylic acid, beta-methylcrotonic acid | |
| CHEBI:37127 | |
| CC(=CC(=O)O)C |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 208-782-7
RTECSNumber : GQ5425000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only