missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylacrylic acid, 98%

Catalog No. AAA1686514
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1686514 25 g
AAA1686522 100 g
AAA1686536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1686514 Supplier Thermo Scientific Chemicals Supplier No. A1686514

CAS: 541-47-9 | C5H8O2 | 100.117 g/mol

Chemical Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chemical Intermediates.

Solubility
Soluble in water, and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from bases.

Chemical Identifiers

CAS 541-47-9
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number MFCD00004366
InChI Key YYPNJNDODFVZLE-UHFFFAOYSA-N
Synonym 3,3-dimethylacrylic acid, senecioic acid, 3-methylcrotonic acid, 3-methyl-2-butenoic acid, 2-butenoic acid, 3-methyl, senecic acid, beta,beta-dimethylacrylic acid, crotonic acid, 3-methyl, beta,beta-dimethacrylic acid, beta-methylcrotonic acid
PubChem CID 10931
ChEBI CHEBI:37127
IUPAC Name 3-methylbut-2-enoic acid
SMILES CC(=CC(=O)O)C

Specifications

Melting Point 66°C to 68°C
Density 0.969
Boiling Point 198°C to 200°C
Flash Point 93°C (199°F)
Quantity 25 g
UN Number UN3261
Beilstein 1720305
Solubility Information Soluble in water,and methanol.
Formula Weight 100.12
Percent Purity 98%
Chemical Name or Material 3,3-Dimethylacrylic acid
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 208-782-7
RTECSNumber GQ5425000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only