missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethyl-3,4-dihydro-2H-thieno[3,4-b][1,4]dioxepine 95.0+%, TCI America™

Catalog No. p-7128765
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5423200MG 200 mg
D54231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5423200MG Supplier TCI America Supplier No. D5423200MG

Chemical Identifiers

CAS 255901-50-9
Molecular Formula C9H12O2S
Molecular Weight (g/mol) 184.25
MDL Number MFCD08705268
InChI Key PUEUIEYRIVFGLS-UHFFFAOYSA-N
Synonym 3,4-(2,2′-Dimethylpropylenedioxy)thiophene
PubChem CID 9834140
IUPAC Name 3,3-dimethyl-2H,3H,4H-thieno[3,4-b][1,4]dioxepine
SMILES CC1(C)COC2=CSC=C2OC1

Specifications

Melting Point 45°C
Quantity 200 mg
Formula Weight 184.25
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,3-Dimethyl-3,4-dihydro-2H-thieno[3,4-b][1,4]dioxepine
TSCA No
Recommended Storage Refrigerator