missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethyl-2-oxobutyric Acid 98.0+%, TCI America™

Catalog No. D36095G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36095G 5 g
D360925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36095G Supplier TCI America Supplier No. D36095G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 815-17-8
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD00154352
InChI Key IAWVHZJZHDSEOC-UHFFFAOYSA-N
Synonym 3,3-dimethyl-2-oxobutyric acid, trimethylpyruvic acid, butanoic acid, 3,3-dimethyl-2-oxo, pyruvic acid, trimethyl, glyoxylic acid, tert-butyl, unii-29cl615clh, 3,3-dimethyl-2-oxo-butanoic acid, butyric acid, 3,3-dimethyl-2-oxo, tert-butylglyoxylic acid, trimethlpyruvic acid
PubChem CID 13150
IUPAC Name 3,3-dimethyl-2-oxobutanoic acid
SMILES CC(C)(C)C(=O)C(=O)O

Specifications

Color Yellow
Boiling Point 80°C
Quantity 5 g
UN Number 3265
Formula Weight 130.14
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 3,3-Dimethyl-2-oxobutyric Acid
EINECSNumber (2)-1503
TSCA Yes