missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethyl-2-butylamine 98.0+%, TCI America™

Catalog No. p-7116990
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mL
25 mL
Catalog No. Quantity
A065610ML 10 mL
A065625ML 25 mL
2 options

Catalog No. A065610ML

Supplier: TCI America A065610ML

Only null left

Chemical Identifiers

CAS 3850-30-4
Molecular Formula C6H16N
Molecular Weight (g/mol) 102.20
MDL Number MFCD00008078
InChI Key DXSUORGKJZADET-YFKPBYRVSA-O
Synonym 1,2,2-trimethylpropylamine, 2-amino-3,3-dimethylbutane, 2-butanamine, 3,3-dimethyl, 3,3-dimethyl-2-aminobutane, 3,3-dimethyl-2-butanamine, 3-amino-2,2-dimethylbutane, propylamine, 1,2,2-trimethyl, 3,3-dimethyl-2-butylamine, 3,3-dimethylbut-2-ylamine, 3,3-dimethyl-2-butyl amine
PubChem CID 520907
IUPAC Name (2S)-3,3-dimethylbutan-2-aminium
SMILES C[C@H]([NH3+])C(C)(C)C

Specifications

Color Colorless
Boiling Point 102°C
Quantity 10 mL
UN Number 2733
Formula Weight 101.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,3-Dimethyl-2-butylamine
EINECSNumber (2)-0133
TSCA Yes