missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethyl-2-butylamine 98.0+%, TCI America™

Catalog No. A065610ML
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A065610ML 10 mL
A065625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A065610ML Supplier TCI America Supplier No. A065610ML

Chemical Identifiers

CAS 3850-30-4
Molecular Formula C6H16N
Molecular Weight (g/mol) 102.20
MDL Number MFCD00008078
InChI Key DXSUORGKJZADET-YFKPBYRVSA-O
Synonym 1,2,2-trimethylpropylamine, 2-amino-3,3-dimethylbutane, 2-butanamine, 3,3-dimethyl, 3,3-dimethyl-2-aminobutane, 3,3-dimethyl-2-butanamine, 3-amino-2,2-dimethylbutane, propylamine, 1,2,2-trimethyl, 3,3-dimethyl-2-butylamine, 3,3-dimethylbut-2-ylamine, 3,3-dimethyl-2-butyl amine
PubChem CID 520907
IUPAC Name (2S)-3,3-dimethylbutan-2-aminium
SMILES C[C@H]([NH3+])C(C)(C)C

Specifications

Color Colorless
Boiling Point 102°C
Quantity 10 mL
UN Number 2733
Formula Weight 101.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,3-Dimethyl-2-butylamine
EINECSNumber (2)-0133
TSCA Yes