missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-3,3-Dimethyl-2-butylamine, 98%

Catalog No. p-7081521
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1153006 5 g
AAA1153014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1153006 Supplier Thermo Scientific Chemicals Supplier No. A1153006

CAS: 3850-30-4 | C6H16N | 102.20 g/mol

(±)-3,3-Dimethyl-2-butylamine is used to produce 1-pyridin-3-yl-3-(1,2,2-trimethyl-propyl)-thiourea. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-3,3-Dimethyl-2-butylamine is used to produce 1-pyridin-3-yl-3-(1,2,2-trimethyl-propyl)-thiourea. It is also used as pharmaceutical intermediates.

Solubility
Fully miscible in water.

Notes
Air Sensitive. Store away from acid chlorides and acid anhydrides.

Chemical Identifiers

CAS 3850-30-4
Molecular Formula C6H16N
Molecular Weight (g/mol) 102.20
MDL Number MFCD00008078
InChI Key DXSUORGKJZADET-YFKPBYRVSA-O
Synonym 1,2,2-trimethylpropylamine, 2-amino-3,3-dimethylbutane, 2-butanamine, 3,3-dimethyl, 3,3-dimethyl-2-aminobutane, 3,3-dimethyl-2-butanamine, 3-amino-2,2-dimethylbutane, propylamine, 1,2,2-trimethyl, 3,3-dimethyl-2-butylamine, 3,3-dimethylbut-2-ylamine, 3,3-dimethyl-2-butyl amine
PubChem CID 520907
IUPAC Name 3,3-dimethylbutan-2-amine
SMILES C[C@H]([NH3+])C(C)(C)C

Specifications

Density 0.755
Quantity 5 g
UN Number UN2733
Sensitivity Air Sensitive
Solubility Information Fully miscible in water.
Formula Weight 101.19
Percent Purity 98%
Chemical Name or Material (±)-3,3-Dimethyl-2-butylamine
Hazard Category H225-H302+H312+H332-H314
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 223-353-4
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only