missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethyl-1-butene, 95%

Catalog No. AC152722500
Change view
Click to view available options
Quantity:
250 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC152722500 250 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC152722500 Supplier Thermo Scientific Chemicals Supplier No. 152722500
Only null left

CAS: 558-37-2 | C6H12 | 84.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 558-37-2
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00008853
InChI Key PKXHXOTZMFCXSH-UHFFFAOYSA-N
Synonym 3,3-dimethyl-1-butene, neohexene, 1-butene, 3,3-dimethyl, tert-butylethylene, tert-butylethene, tert-hexene, trimethylvinylmethane, 3,3-dimethylbutene, 2,2-dimethyl-3-butene, unii-01zb73d2kk
PubChem CID 11210
IUPAC Name 3,3-dimethylbut-1-ene
SMILES CC(C)(C)C=C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point -115.0°C
Color Colorless
Density 0.6500g/mL
Boiling Point 41.0°C
Flash Point −28°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CCH=CH2
Refractive Index 1.3750 to 1.3780
Quantity 250 mL
Beilstein 01, 217
Specific Gravity 0.65
Formula Weight 84.16
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3,3-Dimethyl-1-butene, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes skin irritation.
May be fatal if swallowed and enters airways.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 209-195-9
TSCA TSCA

RUO – Research Use Only