missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3-Dimethyl-1-butanol 95.0+%, TCI America™
Supplier: TCI America D133325ML
Specifications
| 3,3-Dimethyl-1-butanol | |
| Yellow | |
| C6H14O | |
| 25 mL | |
| 3,3-dimethyl-1-butanol, 3,3-dimethylbutanol, 1-butanol, 3,3-dimethyl, neohexanol, unii-543oyd666t, tert-hexyl alcohol, 3,3-dimethyl-butan-1-ol, chembl25029, tert-hexanol, pubchem3096 | |
| CC(C)(C)CCO | |
| 102.18 | |
| 102.18 | |
| Liquid |
| 624-95-3 | |
| 142°C | |
| MFCD00002928 | |
| 2282 | |
| DUXCSEISVMREAX-UHFFFAOYSA-N | |
| 3,3-dimethylbutan-1-ol | |
| 12233 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 624-95-3 | |
| 102.18 | |
| DUXCSEISVMREAX-UHFFFAOYSA-N | |
| 12233 | |
| CC(C)(C)CCO |
| C6H14O | |
| MFCD00002928 | |
| 3,3-dimethyl-1-butanol, 3,3-dimethylbutanol, 1-butanol, 3,3-dimethyl, neohexanol, unii-543oyd666t, tert-hexyl alcohol, 3,3-dimethyl-butan-1-ol, chembl25029, tert-hexanol, pubchem3096 | |
| 3,3-dimethylbutan-1-ol |
Safety and Handling
EINECSNumber : (2)-0217
TSCA : Yes